Learn More
Merck Amicon™ Stirred Cell Maintenance Kits (Stir Bar Assembly, 2 Seals, Tubing kit)
Product Code. 17514715 Shop All Merck Products
Change view
Click to view available options
For Use With (Equipment):
Amicon Stirred Cell (200 mL)
Amicon Stirred Cell (50 mL)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17514715 Amicon Stirred Cell (200 mL) Each
15836432 Amicon Stirred Cell (50 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17514715 Supplier Merck Supplier No. UFSC200MT
 Restricted Item
Estimated Shipment: 09-12-2026
Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.
This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.
TRUSTED_SUSTAINABILITY
Specifications
Description Amicon Stirred Cell 200 mL Maintenance kit
Includes Amicon™ Stirred Cell 200 mL Maintenance kit (stir bar assembly, 2 Ea. seals, tubing kit)
Type Ultrafiltration Equipment
For Use With (Equipment) Amicon Stirred Cell (200 mL)