Learn More
Cytiva Amersham™ Hyperfilm™ ECL
Product Code. 10752067 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (LxW):
12.7 x 17.8 cm (5 x 7 in.)
18 x 24 cm (7.1 x 9.5 in.)
20.3 x 25.4 cm (8 x 10 in.)
Quantity:
100 Sheets/Box
50 Sheets/Box
Unit Size:
100 sheets
50 sheets
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Quantity Unit Size
10752067 12.7 x 17.8 cm (5 x 7 in.) 50 Sheets/Box 50 sheets
10534205 18 x 24 cm (7.1 x 9.5 in.) 100 Sheets/Box 100 sheets
10479214 20.3 x 25.4 cm (8 x 10 in.) 50 Sheets/Box 50 sheets
10607665 18 x 24 cm (7.1 x 9.5 in.) 50 Sheets/Box 50 sheets
10075224 20.3 x 25.4 cm (8 x 10 in.) 100 Sheets/Box 100 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10752067 Supplier Cytiva Supplier No. 28906835
  • ONLINE EXCLUSIVE PRICE
    294.78 EUR
    219.00 EUR / 50 sheets
    Save 75.78 EUR
    25% Off
 Restricted Item
In Stock
Use with all blue and green light chemiluminescent systems. Cytiva Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
  •  Antistatic coating reduces risk of visible artifacts
  • Clear base offers high contrast for easy reading of images
  • Suitable for use with all blue and green light chemiluminescent systems
  • Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
  • Chemiluminescent detection in Western, Southern, and Northern blots
TRUSTED_SUSTAINABILITY
Specifications
Product Type Amersham Hyperfilm ECL
For Use With (Application) Chemiluminescent detection in Western, Southern, and Northern blots.
Quantity 50 Sheets/Box
Content And Storage 4°C
Dimensions (LxW) 12.7 x 17.8 cm (5 x 7 in.)