Learn More
Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs
Product Code. 11738278 Shop All Cytiva Products
Change view
Click to view available options
Label or Dye:
Cy2 bis-Reactive Dye
Cy3 mono-Functional Dye
Cy3.5 mono-Functional Dye
Cy5 Reactive Dye
Cy5 mono-Functional Dye
Cy5.5 bis-Functional Dye
Cy5.5 mono-Functional Dye
Unit Size:
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Label or Dye unitSize
11738278 Cy5.5 mono-Functional Dye Pack of 5
11798268 Cy3.5 mono-Functional Dye Pack of 5
11758268 Cy2 bis-Reactive Dye Pack of 5
11708278 Cy5 Reactive Dye Pack of 5
11728278 Cy5.5 bis-Functional Dye Pack of 5
11718278 Cy5 mono-Functional Dye Pack of 5
10156044 Cy3 mono-Functional Dye Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11738278 Supplier Cytiva Supplier No. PA25501
Use for the most accurate labeling. Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.
    • Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
    • For use in ELISA, immunoprecipitation, and blotting applications.
    • Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.
Order Info
Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For the labelling of biological compounds with Cy5.5 monofunctional dye
Quantity Each
Product Type Cy5.5 Mono-Reactive Dye Pack
Kit Contents Five foil packs each containing dried dye to label 1mg of Protein
Sufficient For Labeling 1mg Protein or 30nM Amino groups
Molecular Weight (g/mol) 1128.41
Content And Storage 2°C to 8°C, protected from light
Label or Dye Cy5.5 mono-Functional Dye
Color Blue
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1