Learn More
Ambroxol hydrochloride, 98%, Thermo Scientific™
49.65 EUR - 170.65 EUR
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15455899
SDS View Documents
Thermo Scientific Alfa Aesar
J63712.03
ONLINE EXCLUSIVE PRICE
54.90 EUR
49.65 EUR / 1g
Save 5.25 EUR
9% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 Restricted Item
15465899
SDS View Documents
Thermo Scientific Alfa Aesar
J63712.06
5g
ONLINE EXCLUSIVE PRICE
189.00 EUR
170.65 EUR / 5g
Save 18.35 EUR
9% Off
View alternative products
Save Up to 
»
Estimated Shipment: 14-09-2026
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Ambroxol hydrochloride is a medication indicated to alleviate chest congestion associated with conditions that include bronchitis, pneumonia, bronchospasm asthma, cough, and allergy. Preclinically, ambroxol, the active ingredient of Mucosolvan, has been shown to increase respiratory tract secretion. It enhances pulmonary surfactant production and stimulates ciliary activity.
Solubility
Slightly soluble in water (∼5 mg/ml) or ethanol (∼5 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.