missing translation for 'onlineSavingsMsg'
Learn More

Ambroxol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30463212
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463212 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463212 Supplier LoGiCal Supplier No. LGCAMP0005.0001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ambroxol hydrochloride
CAS 23828-92-4
Analyte or Component Names Ambroxol hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C13 H18 Br2 N2 O . Cl H
Certifications/Compliance ISO 17034
Synonym Cyclohexanol, 4-[[(2-amino-3,5-dibromophenyl)methyl]amino]-, hydrochloride (1:1), trans-, Cyclohexanol, 4-[(2-amino-3,5-dibromobenzyl)amino]-, hydrochloride, trans- (8CI), Cyclohexanol, 4-[[(2-amino-3,5-dibromophenyl)methyl]amino]-, monohydrochloride, trans- (9CI), Abramen, Ambril, Ambrodil, Ambron, Ambroxol hydrochloride, Bronchopront, Coughnol, Duramucal, Fluibron, Fluixol, Frenopect, Lindoxyl, Motosol, Mucasan, Muco-Burg, Mucoclear, Mucofar, Mucolear, Mucolin, Mucolite, Mucosal, Mucosolvan, Mucovent, N-(trans-4-Hydroxycyclohexyl)-2-amino-3,5-dibromobenzylamine hydrochloride, Pect, Solvolan, Stas-Hustenloser, Surbronc, Surfactal, trans-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol hydrochloride
InChI Formula InChI=1S/C13H18Br2N2O.ClH/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10;/h5-6,10-11,17-18H,1-4,7,16H2;1H/t10-,11-;
Solution Type Single Solution
SMILES Cl.Nc1c(Br)cc(Br)cc1CN[C@@H]2CC[C@@H](O)CC2
IUPAC Name 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 414.56
Quantity 1 mL
Formula Weight 411.9553 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.