Learn More
Invitrogen™ RNaseAlert™ Lab Test Kit
Product Code. 10249204
Change view
Click to view available options
Quantity:
25 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10249204 25 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10249204 Supplier Invitrogen™ Supplier No. AM1964
Accelerate Your Research With Exclusive Life Science Offers
Use promo code 28141
View all promotions
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time.
Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time. Suitable for testing small sample numbers and can be used to ensure that solutions, tubes, tips, etc. are RNase-free; the kit contains sufficient reagents for 25 reactions.
  • Detects as little as 3.5 x 10-7 units (∼0.5 pg) of RNase A, with a fluorometer or by eye
  • Simple, straightforward 30 min. assay
  • Monitor RNase activity in column fractions during protein purification
  • Use with DNaseAlert™ to quantitatively detect both RNases and DNases in the same sample
TRUSTED_SUSTAINABILITY
Specifications
Format Lyophilized
Content And Storage Store Florescent Substrate, 10X RNaseAlert Lab Test Buffer, and RNase A at -20°C.
Store RnaseZap™ and nuclease-free water at room temperature.
Shipping Condition Dry Ice
Quantity 25 reactions
Reagent Type RNase Control Reagent
Product Line Ambion, RNaseAlert
What is the concentration of RNase A in the RNaseAlert Lab Test Kit (Cat. No. AM1964)?
The concentration of RNase A in the RNaseAlert Lab Test Kit is proprietary.