missing translation for 'onlineSavingsMsg'
Learn More

Amaranth (>85%), TRC

Product Code. 30135648
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135648 50 g 50g
30134909 25 g 25g
17923565 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135648 Supplier Toronto Research Chemicals Supplier No. TRCA57598850G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915-67-3
Molecular Formula C20H11N2Na3O10S3, C20 H11 N2 O10 S3 . 3 Na
Synonym 2,7-Naphthalenedisulfonic acid, 3-hydroxy-4-[(4-sulfo-1-naphthalenyl)azo]-, trisodium salt (9CI)∣Bordeaux S (6CI)∣C.I. Acid Red 27 (7CI)∣C.I. Acid Red 27, trisodium salt (8CI)∣1-(4-Sulfo-1-naphthylazo)-2-naphthol-3,6-disulfonic acid trisodium salt∣1302 Red∣1508 Red∣2-Hydroxy-1,1'-azonaphthalene-3,6,4'-trisulfonic acid trisodium salt∣Acid Amaranth∣Acid Amaranth I∣Acid Amaranth J∣Acid Amaranth N∣Acid Leather Red I 2BW∣Acid Leather Rubine S∣Acid Red 27∣Aizen Amaranth∣Amacid Amaranth∣Amaranth∣Amaranth (dye)∣Amaranth 307018∣Amaranth 36010∣Amaranth 85∣Amaranth A∣Amaranth B∣Amaranth BPC∣Amaranth Extra∣Amaranth Lake∣Amaranth R∣Amaranth S∣Amaranth S Specially Pure∣Amaranth USP∣Amaranth WD∣Amaranth red∣Amaranthe∣Azo Red R∣Azo Rubine S-FQ∣Azo Rubine SF∣Azo Ruby S∣Azorubin S∣Basovit Red 470E∣Bordeaux S Extra Conc. A Export∣Bordeaux S Extra Pure A∣Borunil Red A-B∣C.I. 16185∣C.I. Food Red 9∣Canacert Amaranth∣Carmine red E 123∣Certicol Amaranth S∣Cogilor Red 318.11∣Cranberry Red∣D & C Red 2∣D and C Red No. 2∣D&C Red No. 2∣Daishiki Amaranth∣Dolkwal Amaranth∣Dye Red Raspberry∣E 123∣Edicol Amaranth∣Edicol Supra Amaranth A∣Edicol Supra Amaranth AS∣Euorcert Allura 311801∣Eurocert Amaranth∣FD and C Red No. 2∣FD&C Red 2∣Fast Red∣Food Red 2∣Food Red 9∣Food Red No. 2∣Fruit Red A Geigy∣HD Amaranth B∣HD Amaranth Supra∣Hexacert Red No. 2∣Hexacol Amaranth B Extra∣Hidacid Amaranth∣Hidacid Amaranth 21-6008∣Hispacid Red AM∣Japan Food Red No. 2∣Japan Red 2∣Japan Red No. 2∣Java Amaranth∣KCA Foodcol Amaranth A∣Kayaku Amaranth∣Kayaku Food Colour Red No. 2∣Kiton Rubine S∣L Red 3∣L Red Z 3050∣Lissamine Amaranth AC∣Maple Amaranth∣Naphthol Red LZS∣Naphthol Red S∣Naphthol Red S Conc. Specially Pure∣Naphthol Red S Specially Pure∣Naphthol Red SI∣Necol Amaranth∣Neklacid Red A∣PuriColor Red ARE 27∣Rakuto Amaranth∣Red No. 2∣Red dye no. 2∣Ritacid Red G∣Rouge Solid E∣S-Azo Rubine∣San-ei Amaranth∣Sandal Amaranth I∣Shikiso Amaranth∣Sicomet Amaranth∣Sicomet Amaranth 85E123∣Solar Red O∣Takaoka Amaranth∣Tertracid Red A∣Toyo Amaranth∣Triacid Amaranth A∣Trisodium salt of 1-(4-sulfo-1-naphthylazo)-2-naphthol-3,6-disulfonic acid∣Usacert Red No. 2∣Victoria Rubine O∣Victory Scarlet∣Water Red 176574∣Whortleberry Red∣Wool Bordeaux 6RK∣Wool Red 40F∣Xiancai Red

Specifications

Format Neat
Formula Weight 603.9269
InChI Key VOBHRQFELWTZFS-DXVNCNPQSA-K
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C(=O)\C2=N\NC1=C2C=CC=CC2=C(C=C1)S([O-])(=O)=O)S([O-])(=O)=O
IUPAC Name trisodium (4E)-3-oxo-4-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Quantity 50 g
Molecular Weight (g/mol) 604.46
Shipping Condition Room Temperature
Solution Type Neat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.