Learn More
MedChemExpress Amaranth
Specifications
Specifications
| CAS | 915-67-3 |
| Molecular Weight (g/mol) | 604.47 g/mol |
| Quantity | 1 g |
| Shipping Condition | Room temperature |
| Storage Requirements | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Solubility | DMSO : 25 mg/mL (41.36 mM; Need ultrasonic) ∣ H2O : 6.67 mg/mL (11.03 mM; Need ultrasonic) |
| Synonym | Acid Red 27 ∣ Azorubin S ∣ FD & C Red Dye No. 2 |
| Target | HBV |
| Product Type | Dye Reagents |