missing translation for 'onlineSavingsMsg'
Learn More

α-Amanitin, MedChemExpress

Product Code. 16490278
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
2 mg
5 mg
Unit Size:
1mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490278 5 mg 5mg
16401839 1 mg 1mg
16400326 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490278 Supplier Medchem Express Supplier No. HY19610
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress α-Amanitin is the principal toxin of several deadly poisonous mushrooms, exerting its toxic function by inhibiting RNA-polymerase II.

α-Amanitin is the principal toxin of several deadly poisonous mushrooms, exerting its toxic function by inhibiting RNA-polymerase II.

Specifications

Chemical Name or Material α-Amanitin
CAS 23109-05-9
Color Light Brown
Purity Grade Notes Research
Molecular Formula C39H54N10O14S
Quantity 5 mg
Synonym α-Amatoxin
Solubility Information H2O : 100 mg/mL (108.82 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (54.41 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C[C@@](NC(CNC([C@@]([C@@H](C)CC)([H])N1)=O)=O)([H])C(N[C@@](C(N(C[C@H](O)C2)[C@]2([H])C3=O)=O)([H])CC(N)=O)=O)C4=C(C[C@](C(NCC1=O)=O)([H])NC([C@](N3)([H])[C@@H](C)[C@@H](O)CO)=O)C5=CC=C(O)C=C5N4
Molecular Weight (g/mol) 918.97
Formula Weight 918.97
Percent Purity 97.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.