Learn More
Tocris Bioscience™ AM 580
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
TB0760-RMU is an Ancillary Material Grade version of product AM 580 (Cat. No. 0760). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
Find out more about our Ancillary Material Grade product range.
Specifications
Specifications
| Chemical Name or Material | 4-[[(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)carbonyl]amino]benzoic acid |
| CAS | 102121-60-8 |
| Quantity | 10 mg |
| Target | Retinoic Acid Receptor Agonists |
| Molecular Formula | C22H25NO3 |
| Purity | 0.98 |