Specifications
| Material | Aluminum |
| Max. Temperature (Metric) | 640°C |
| Quantity | 1Pack |
| Capacity (Metric) | 40 μL |
| Diameter (Metric) Outer | 6mm |
| Height (Metric) | 1.6mm |
| Product Type | Standard Crucible |