Learn More
Cole-Parmer™ Aluminum Smooth Wall Weighing Dishes
Product Code. 11735459 Shop All Cole Parmer Products
Change view
Click to view available options
Capacity (Metric):
80 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11735459 80 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11735459 Supplier Cole-Parmer™ Supplier No. 0101829
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 30-12-2026
Use these dishes for a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical and pharmaceutical. Cole-Parmer™ Aluminum Smooth Wall Weighing Dishes are ideal for storage, dispensing, general purpose sampling and testing.
  • Smooth-sided, flat bottom, multipurpose “Utility” dishes
  • Oil-free and non-coated, no tab
  • Dish contour facilitates stacking and dispensing
  • Disposable
Specifications
Bottom Shape Flat
Capacity (Metric) 80 mL
Disposable Single-use
Material Aluminum
Shape Flat
Type Weighing Dish
For Use With (Application) For storage, dispensing, general purpose sampling and testing
For Use With (Equipment) Weighing, Moisture Testing, Wastewater Analysis, Chemical, and Pharmaceutical
Width (Metric) 1.7 cm
Depth (Metric) 7.3 cm
Diameter (English) 2.875 in.
Diameter (Metric) 1.7 cm