missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kern Sohn Aluminum Protective Box
Description
- Box for individual weights (cylindrical)
- For safe storage and transportation under harsh industrial conditions
Specifications
Specifications
| Product Type | Case |
| Classification | E1 - F2 |
| Case Material | Aluminum |
| For Use With (Equipment) | Button/Compact Weights |
| Holds | 50 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?