Learn More
Korff™ Aluminum Professional Rolls
Product Code. 10533282
Change view
Click to view available options
Width (Metric):
300mm
450mm
600mm
Unit Size:
100m
150m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
10533282 300mm 150m
10322232 600mm 100m
10730193 450mm 150m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10533282 Supplier Korff™ Supplier No. 60100
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Estimated Shipment: 30-11-2026