Learn More
Schneider Gerd™ Aluminum Needle Holder
Product Code. 10545524
Change view
Click to view available options
Material:
Aluminum
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10545524 Aluminum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10545524 Supplier Schneider Gerd™ Supplier No. 262
Only null left
Add to basket
Add to basket
Use this needle holder for dissection applications. Schneider Gerd™ Aluminum Needle Holders feature a plastic handle for a secure grip.
  • Aluminum Needle Holder
  • Available in a variety of colors and sizes
Specifications
Product Type Needle Holder
Material Aluminum
For Use With (Equipment) Dissection needles
Length (Metric) 240 mm