Learn More
Fisherbrand™ Aluminum Labjacks
Product Code. 12484060 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
128 to 470 mm
36 to 78 mm
43 to 140 mm
57 to 290 mm
60 to 290 mm
Length (Metric) Base:
50 mm
100 mm
150 mm
200 mm
320 mm
Width (Metric) Base:
40 mm
100 mm
150 mm
200 mm
260 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Length (Metric) Base Width (Metric) Base Unit Size
12484060 57 to 290 mm 200 mm 200 mm Each
11775386 43 to 140 mm 100 mm 100 mm Each
11825091 60 to 290 mm 150 mm 150 mm Each
11735396 36 to 78 mm 50 mm 40 mm Each
12862527 128 to 470 mm 320 mm 260 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12484060
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
Provides for precise height adjustment with the turn of a screw. Fisherbrand™ Aluminum Labjacks are manufactured to close tolerances with a stainless steel operating screw.
  • Blue anodized finish
  • Completely stable and free from rocking movements
  • Top and bottom plates are specially designed aluminum extrusion giving exceptional rigid and robust support
Specifications
Material Aluminum
Height (Metric) 57 to 290 mm
Length (Metric) Base 200 mm
Width (Metric) Base 200 mm
Includes With stainless steel operating screw
Product Type Lab Jack
Quantity 1 Ea.