Learn More
Welch™ Aluminum KF Inner Centering with Buna N O-ring
Product Code. 10762152 Shop All Welch Ilmvac Products
Change view
Click to view available options
Diameter (Metric) Outside:
16 mm
25 mm
40 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Unit Size
10762152 16 mm Each
10640153 25 mm Each
10596292 40 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10762152 Supplier Welch™ Supplier No. 701081
  • ONLINE EXCLUSIVE PRICE
    6.55 EUR
    6.20 EUR / Pack of 1
    Save 0.35 EUR
    5% Off
Estimated Shipment: 04-01-2027
Specifications
Diameter (Metric) Outside 16 mm
For Use With Separator 16 DN
Material Aluminum
Quantity 1 Each
Type KF Inner Centering