Specifications
| Material | Aluminum |
| Quantity | 1 Ea. |
| Style | with Up Arrow |
| For Use With (Application) | for Pipe Markers and Safety Signs |
| Height (Metric) | 297 mm |
| Max. Temperature (Metric) | 120°C |
| Width (Metric) | 148 mm |
| Displayed Text | Exit and emergency exit |
| Product Type | Evacuation Panel Safety Sign |
| Color | White on Green |