missing translation for 'onlineSavingsMsg'
Learn More

Aluminium Nitrate Nonahydrate, Supelco™

Product Code. 15896940
Change view
Click to view available options
Quantity:
1 kg
50 kg
Unit Size:
1kg
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896940 50 kg 50kg
15844020 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15896940 Supplier Supelco Supplier No. 1010869050
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-27-2
Grade Research
Molecular Formula Al(NO3)3 · 9 H2O
Molecular Weight (g/mol) 375.13

Specifications

Melting Point 73°C
Quantity 50 kg
MDL Number MFCD00149132
Synonym Aluminium nitrate nonahydrate, Aluminium nitrate nonahydrate
Density 1.72 g/cm3 at 20°C
Chemical Name or Material Aluminium Nitrate Nonahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.