Learn More
Aluminium Ammonium Sulfate Dodecahydrate, Supelco™
Specifications
Specifications
| Boiling Point | 200°C/1013 hPa |
| Melting Point | 93°C |
| CAS | 7784-26-1 |
| Quantity | 500 g |
| Molecular Formula | NH4Al(SO4)2 · 12 H2O |
| MDL Number | MFCD00149958 |
| Synonym | Aluminium ammonium sulfate dodecahydrate, Alum ammonium, Ammonium alum, Aluminium ammonium sulfate dodecahydrate |
| Molecular Weight (g/mol) | 453.33 |
| Grade | Research |
| Chemical Name or Material | Aluminium Ammonium Sulfate Dodecahydrate |