missing translation for 'onlineSavingsMsg'
Learn More

Altrenogest, MedChemExpress

Product Code. 16462166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462166 500 mg 500mg
16418356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462166 Supplier Medchem Express Supplier No. HYB0521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Altrenogest (Allyltrenbolone) is a progestogen structurally related to veterinary steroid trenbolone.

Altrenogest (Allyltrenbolone) is a progestogen structurally related to veterinary steroid trenbolone.

Specifications

Chemical Name or Material Altrenogest
Physical Form Solid
CAS 850-52-2
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C21H26O2
Synonym Allyltrenbolone RU2267
Solubility Information DMSO : ≥ 100 mg/mL (322.13 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@]([C@]1(O)CC=C)(C=C2)[C@](CC1)([H])[C@@](CC3)([H])C2=C(CC4)C3=CC4=O
Molecular Weight (g/mol) 310.43
Formula Weight 310.43
Percent Purity 99.8%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.