missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alprenolol Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Alprenolol hydrochloride |
| CAS | 13707-88-5 |
| Analyte or Component Names | Alprenolol hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H23 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Alprenolol hydrochloride, (2RS)-1-[(1-Methylethyl)amino]-3-[2-(prop-2-enyl)phenoxy]propan-2-ol hydrochloride, 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propen-1-yl)phenoxy]-, hydrochloride (1:1), 2-Propanol, 1-(o-allylphenoxy)-3-(isopropylamino)-, hydrochloride (8CI), 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propenyl)phenoxy]-, hydrochloride (9CI), (±)-1-(o-Allylphenoxy)-3-isopropylamino-2-propanol hydrochloride, (±)-Alprenolol hydrochloride, 1-(2-Allylphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(o-Allylphenoxy)-3-(isopropylamino)-2-propanol hydrochloride, Alfeprol, Alprenolol hydrochloride, Apllobal, Applobal, Aprobal, Aptin, Aptine, Aptol Duriles, Betaptin, DL-1-(o-Allylphenoxy)-3-isopropylamino-2-propanol hydrochloride, Dimacor, Gubernal, H 56/28, Regletin, Yobir, dl-Alprenolol chloride |
| InChI Formula | InChI=1S/C15H23NO2.ClH/c1-4-7-13-8-5-6-9-15(13)18-11-14(17)10-16-12(2)3;/h4-6,8-9,12,14,16-17H,1,7,10-11H2,2-3H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CC(C)NCC(O)COc1ccccc1CC=C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction