Learn More
Ansell™ AlphaTec™MICROCHEM™4000 Model 215 Apron with Sleeves
Product Code. 15885342 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15885342 2X-Large Pack of 20
15855342 Small Pack of 20
15865342 Medium Pack of 20
15875342 Large Pack of 20
15895342 3X-Large Pack of 20
15805352 4X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15885342 Supplier Ansell™ Supplier No. GR40T0021505
  • ONLINE EXCLUSIVE PRICE
    1590.80 EUR
    1040.46 EUR / Pack of 20
    Save 550.34 EUR
    34% Off
Estimated Shipment: 08-01-2027
MICROCHEM™ 4000 is designed to provide anexceptional barrier against organic and inorganicchemicals and biological agents
  • Velcro fastening to neck
  • Tie fastening at the waist
  • Double cuff with knitted
  • inner cuff
  • Welded and taped seams
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size 2X-Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Disposable Limited-use
Closure Type Hook and Loop, Waist Ties
Sterility Non-sterile
Cuff Style Double Inner Knit
Latex Free Yes
No. of Pockets None
Resistance Abrasions, acids, anti-static, bloodborne pathogens, chemicals
Certifications/Compliance EN 14605: 2005 +A1:2009, Type PB (3); EN 14605: 2005 +A1:2009, Type PB (4); EN14126: 2003, Types PB(3)-B, PB (4)-B
Style Alphatec™ 4000, Model 215
Seam Type Welded and Taped