Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Protective Coveralls
- Chemical Resistant Coveralls
- Ansell AlphaTec™ 4000
Ansell AlphaTec™ 4000
Product Code. 17110675
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
G40T103
G40T111
G40T121
G40T125B
G40T125D
G40T132
G40T151
G40T151B
G40T162
Unit Size:
Pack of 10
Pack of 12
Pack of 8
37 product options available for selection
| Product Code. | Size | Style | Unit Size |
|---|---|---|---|
| 17110675 | Large | G40T151 | Pack of 8 |
| 17110645 | X-Small | G40T111 | Pack of 10 |
| 17120655 | Medium | G40T125D | Pack of 8 |
| 17160635 | Medium | G40T103 | Pack of 10 |
| 17170635 | Large | G40T103 | Pack of 10 |
| 17180635 | X-Large | G40T103 | Pack of 10 |
| 17190635 | 2X-Large | G40T103 | Pack of 10 |
| 17100645 | 3X-Large | G40T103 | Pack of 10 |
| 17190645 | Small | G40T125B | Pack of 8 |
| 17110655 | Medium | G40T125B | Pack of 8 |
| 17130655 | Large | G40T125B | Pack of 8 |
| 17150655 | X-Large | G40T125B | Pack of 8 |
| 17170655 | 2X-Large | G40T125B | Pack of 8 |
| 17190655 | 3X-Large | G40T125B | Pack of 8 |
| 17100655 | Small | G40T125D | Pack of 8 |
| 17120645 | Small | G40T121 | Pack of 10 |
| 17140655 | Large | G40T125D | Pack of 8 |
| 17160655 | X-Large | G40T125D | Pack of 8 |
| 17180655 | 2X-Large | G40T125D | Pack of 8 |
| 17100665 | 3X-Large | G40T125D | Pack of 8 |
| 17130665 | Small | G40T132 | Pack of 12 |
| 17140665 | Medium | G40T132 | Pack of 12 |
| 17150665 | Large | G40T132 | Pack of 12 |
| 17160665 | X-Large | G40T132 | Pack of 12 |
| 17170665 | 2X-Large | G40T132 | Pack of 12 |
| 17180665 | 3X-Large | G40T132 | Pack of 12 |
| 17100675 | Medium | G40T151 | Pack of 8 |
| 17120675 | X-Large | G40T151 | Pack of 8 |
| 17130675 | 2X-Large | G40T151 | Pack of 8 |
| 17140675 | 3X-Large | G40T151 | Pack of 8 |
| 17190665 | Small | G40T151B | Pack of 8 |
| 17150675 | Small | G40T162 | Pack of 10 |
| 17160675 | Medium | G40T162 | Pack of 10 |
| 17170675 | Large | G40T162 | Pack of 10 |
| 17180675 | X-Large | G40T162 | Pack of 10 |
| 17190675 | 2X-Large | G40T162 | Pack of 10 |
| 17100685 | 3X-Large | G40T162 | Pack of 10 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 37 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Beschreibung
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect, typical innovative features include dual zip systems and double cuffs
- Welded and taped seams, our highest barrier to liquids and particulates
- Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
- Anti-static
Spezifikation
Spezifikation
| Size | Large |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Green |
| Seam Type | Welded, Taped |
| Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003, NFPA 1992 |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
| Weight (Metric) | 103 gsm |
| Style | G40T151 |
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?