Description
- Composed of protective multi-layer barrier fabric, AlphaTec™ 3000 Overshoes Ultrasonically Welded Taped Model 400 affords superb resistance against inorganic chemicals and biological hazards
- Bright yellow in color, these overshoes not only offer chemical resistance but also high visibility in dark environments to boost worker safety
- In addition, they are among the most comfortable, lightweight and robust protective footwear on the market
Specifications
Specifications
| Size | One Size |
| Material | Multi-layer Barrier Laminate Fabric |
| Latex Free | Yes |
| Color | Yellow |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Opening |
| Sole Type | Standard |
| Certifications/Compliance (Europe) | CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126 |
| Resistance | Chemical-resistant, Bloodborne Pathogens, Light Chemical Splash, Airborne Solid Particulates |
| Show More |
Product Title
Haben Sie Verbesserungsvorschläge?