Learn More
Ansell™ AlphaTec™ 3000 Cape Hood with Visor Ultrasonically Welded and Taped
Product Code. 31114789 Shop All Ansell Products
Change view
Click to view available options
Closure Type:
Pull-over
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type
31114789 Pull-over
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 31114789 Supplier Ansell™ Supplier No. YE30T0050800
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
Estimated Shipment: 24-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Protective multi-layer barrier fabric effective against numerous chemicals, featuring lightweight and durable
  • AlphaTec™ 3000 Cape Hood with Visor Welded and Taped - Model 508 is designed with a protective multi-layer barrier fabric, which is extremely effective against a range of harmful inorganic chemicals and biological hazards
  • With their highly visible bright yellow color, these chemical protective garments reinforce the safety of workers
  • Lightweight and durable, this protective overgarment ensures sustained comfort and durable coverage for its wearers
  • As it is low linting, this protective garment minimizes the risk of product contamination
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hood with Visor
Size One Size
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Closure Type Pull-over
Disposable Single-use
Sterility Non-sterile
Certifications/Compliance CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126)