Learn More
Ansell™ AlphaTec™ 3000 Apron - Model 213
Product Code. 31114774 Shop All Ansell Products
Change view
Click to view available options
Closure Type:
Ties
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type
31114774 Ties
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31114774 Supplier Ansell™ Supplier No. YE30T0021300
Estimated Shipment: 04-12-2026
Protective multi-layer barrier, protection against numerous chemicals with Lightweight and durable material
  • This AlphaTec™ chemical-protective apron features a multi-layer fabric that protects against various inorganic chemicals and biological hazards
  • Bright yellow color, ensures highly noticeable personal protection, enhancing workplace safety and safeguarding worker welfare
  • An extremely light protective apron, the AlphaTec™ 3000 Apron - Model 213 remains highly comfortable, even with extended wear
  • Its multi-layer fabric construction also means this chemical-resistant apron withstands wear and tear
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Universal
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Closure Type Ties
Sterility Non-sterile
Latex Free Yes
No. of Pockets None
Seam Type Seamless
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126)