missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Alphacel is a polysaccharide composed of long chains of β(1,4) linked D-glucose units.
- Cellulose is used in a wide range of applications that require a structural back-bone.
- It is used in the preparation of chromatographic structural media, in gels for processes such as electrophoresis and ionic separations and as a substrate to identify and study cellulosic systems.
Specifications
Specifications
| Color | White |
| pH | 5.5 to 7.0 |
| Physical Form | Powder |
| Quantity | 25 lb. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?