Learn More
Alpha-Zingiberene, TRC
Product Code. 30454710
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454710 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30454710 Supplier Toronto Research Chemicals Supplier No. TRCZ4399805MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material alpha-Zingiberene
CAS 495-60-3
Molecular Formula C15 H24
Quantity 5 mg
Synonym 1,3-Cyclohexadiene','5-[(1S)-1,5-dimethyl-4-hexen-1-yl]-2-methyl-','(5R)-, 1,3-Cyclohexadiene','5-(1,5-dimethyl-4-hexenyl)-2-methyl-','[S-(R*,S*)]-, 1,3-Cyclohexadiene','5-[(1S)-1,5-dimethyl-4-hexenyl]-2-methyl-','(5R)- (9CI), Zingiberene (7CI), (5R)-5-[(1S)-1,5-Dimethyl-4-hexen-1-yl]-2-methyl-1,3-cyclohexadiene, (-)-Zingiberene, l-Zingiberene, α-Sesquiphellandrene, α-Zingiberene
InChI Formula InChI=1S/C15H24/c1-12(2)6-5-7-14(4)15-10-8-13(3)9-11-15/h6,8-10,14-15H,5,7,11H2,1-4H3/t14-,15+/m0/s1
SMILES C[C@@H](CCC=C(C)C)[C@H]1CC=C(C)C=C1
IUPAC Name (5R)-2-methyl-5-[(2S)-6-methylhept-5-en-2-yl]cyclohexa-1,3-diene
Molecular Weight (g/mol) 204.35
Formula Weight 204.1878