Learn More
Skytec™ Alpha Sentinel Premium Reusable Full Face Mask
Product Code. 30242954
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30242954 Small Each
30242980 Medium Each
30242985 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30242954 Supplier Skytec™ Supplier No. ASRAS0002BD
  • ONLINE EXCLUSIVE PRICE
    237.43 EUR
    150.37 EUR / Pack of 1
    Save 87.06 EUR
    36% Off
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Alpha Sentinel Full Face Masks are ergonomically designed for optimum protection, comfort and PPE compatibility.
  • The five-point adjustable head harness removes the feeling of a downward pull, ensuring a balanced, comfortable fit
  • Well-balanced, lightweight (600 g)
  • Available in three sizes for a more tailored fit, increasing respirator/fit factor
  • Raised visor bridge profile protects the visor from scratches when not in use
  • Visor and filter position provides panoramic field of view without any obstructions
  • Low profile shape provides exceptional compatibility with other PPE
  • The Alpha Sentinel filters are fitted with a simple connection which easily clicks into position
  • The large exhalation valve reduces heat and moisture build-up, providing low breathing resistance
  • Easily cleaned and maintained, with spare parts available
  • Tested to EN136:1998 Class 2, ensuring greater robustness and higher flame retardancy
  • Supplied with user seal check covers, which double as particulate filter holder for combination filter
  • Optical insert available
  • Quantitative face fit adapter available
  • The harness is designed to prevent over-rotation and tangling
Specifications
Certifications/Compliance CE , UKCA , EN 136:1998 Class 2
Certifications (EN 136) Class 2
Color Black
Filter Type HEPA, Combination Organic Vapor/Acid Gas/HEPA, Multi-Gas/HEPA
Product Type Full Facepiece Respirator
Size Small
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
Model Alpha Sentinel Full Face Mask