missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Skytec™ Alpha Sentinel A2 Filters - Pair Premium Alpha Sentinel Filters
Description
- Gas filters are snowstorm-filled to optimise space and volume, resulting in a smaller lightweight casing that limits obstruction of view when fitted
- Particulate P3S and P3H contain pleated filter media, providing more surface area for reduced breathing resistance
- Sentinel filters are fitted with a simple connection that easily clicks into position
- All filter combinations are lightweight and can be used on both Full Face and Half Mask
- The P3P Filter is tested to 'R', allowing the use for more than one shift
Specifications
Specifications
| Certifications/Compliance | CE , UKCA , EN 14387:2021 A,2 |
| Connection Type | Bayonet |
| Color | Black |
| Filter Type | Combination |
| Product Type | Filter |
| With Exhalation Valve | No |
| For Use With (Equipment) | Alpha Sentinel Full or Half Mask |
| Quantity | 4/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?