Learn More
alpha-Naphthyl Glycidyl Ether, 90%, TRC
Product Code. 30480758
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480758 2.5 g 2.5g
30468196 1 g -
30469022 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30480758 Supplier Toronto Research Chemicals Supplier No. TRCN3784002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2461-42-9
Molecular Formula C13 H12 O2
Molecular Weight (g/mol) 200.23
Synonym 1-(1-Naphthyloxy)-2,3-epoxypropane
IUPAC Name 2-(naphthalen-1-yloxymethyl)oxirane
SMILES C(Oc1cccc2ccccc12)C3CO3
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C13H12O2/c1-2-6-12-10(4-1)5-3-7-13(12)15-9-11-8-14-11/h1-7,11H,8-9H2
Formula Weight 200.0837
Chemical Name or Material 1-(1-Naphthyloxy)-2,3-epoxypropane