missing translation for 'onlineSavingsMsg'
Learn More

alpha-Monothioglycerol, 99.1%, For GC analysis, MP Biomedicals™

Product Code. 11434921 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434921 250 mL 250mL
11404921 25 mL 25mL
11414921 50 mL 50mL
11424921 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11434921 Supplier MP Biomedicals Supplier No. 0215572383
  • ONLINE EXCLUSIVE PRICE
    376.00 EUR
    358.05 EUR / 250mL
    Save 17.95 EUR
    4% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 96-27-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
Synonym 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol
PubChem CID 7291
ChEBI CHEBI:74537
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

Color Colorless
Quantity 250 mL
Formula Weight 108.2
Percent Purity 0.991
Physical Form Liquid
Chemical Name or Material α-Monothioglycerol, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.