missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Methylstyrene-d10, TRC

Product Code. 30482091
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482091 2 mg 2mg
30469720 1 mg 1mg
30470378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30482091 Supplier Toronto Research Chemicals Supplier No. TRCM3253962MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10362-82-0
Molecular Formula C9 D10
Molecular Weight (g/mol) 128.237
Synonym Styrene-β,β,2,3,4,5,6-d7, α-methyl-d3- (8CI), α-(Methyl-d3)styrene-β,β,2,3,4,5,6-d7, [1-(Methyl-d3)ethenyl-2,2-d2]benzene-2,3,4,5,6-d5, α-Methylstyrene-d10, α-Methylstyrene-D10
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,1,3,3,3-pentadeuterioprop-1-en-2-yl)benzene
SMILES [2H]C(=C(c1c([2H])c([2H])c([2H])c([2H])c1[2H])C([2H])([2H])[2H])[2H]

Specifications

Quantity 2 mg
InChI Formula InChI=1S/C9H10/c1-8(2)9-6-4-3-5-7-9/h3-7H,1H2,2H3/i1D2,2D3,3D,4D,5D,6D,7D
Formula Weight 128.141
Chemical Name or Material alpha-Methylstyrene-d10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.