missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-D-Ribose-1-phosphate bis-Cyclohexylammonium Salt, ≥96%, MP Biomedicals™
Shop All MP Biomedicals ProductsSpecifications
Specifications
| CAS | 58459-37-3 |
| Color | White |
| Molecular Formula | C17H37N2O8P |
| Quantity | 5 mg |
| Synonym | alpha-d-ribofuranose-1-phosphate, d-ribofuranose 1-dihydrogen phosphate , compound with cyclohexylamine 1:2, bis cyclohexylamine d-ribose 1-phosphate, d-ribose 1-phosphate,biscyclohexylammonium salt, d-ribose 1-phosphate; bis cyclohexylamine, d-ribose 1-phosphate bis cyclohexylammonium salt tlc, cyclohexanamine hemi 3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl phosphate, cyclohexanamine; 3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl dihydrogen phosphate |
| InChI Key | SSHMAENVNUVVFI-VNISEONSSA-N |
| SMILES | C1CCC(CC1)N.C1CCC(CC1)N.C(C1C(C(C(O1)OP(=O)(O)O)O)O)O |
| IUPAC Name | cyclohexanamine;[(3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl] dihydrogen phosphate |
| Molecular Weight (g/mol) | 428.463 |
| PubChem CID | 6453623 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?