missing translation for 'onlineSavingsMsg'
Learn More

Alpha-D-Glucose 1-Phosphate Disodium Salt Tetrahydrate, TRC

Product Code. 30178944
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178944 5 g 5g
30179532 10 g 10g
30179497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30178944 Supplier Toronto Research Chemicals Supplier No. TRCG5968185G
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56401-20-8
Molecular Formula C6H11Na2O9P . 4(H2O)
Molecular Weight (g/mol) 260.14
Synonym Phocytan Tetrahydrate, alpha-D-Glucopyranosyl phosphate Disodium Salt;alpha-D-Glucose 1-(Dihydrogen Phosphate) Disodium Saltalpha-D-Glucose 1-Phosphate Disodium Salt
IUPAC Name disodium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] phosphate
SMILES O[C@@H]1[C@H]([C@@H]([C@H](O[C@@H]1OP(O[Na])(O[Na])=O)CO)O)O.[4H2O]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H13O9P.2Na/c7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13;;/h2-10H,1H2,(H2,11,12,13);;/q;2*+1/p-2/t2-,3-,4+,5-,6-;;/m1../s1
Formula Weight 303.9936
Chemical Name or Material alpha-D-Glucose 1-Phosphate Disodium Salt Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.