Learn More
Alfa Aesar™ alpha-D-Glucose-1-phosphate dipotassium salt hydrate, 98%, Thermo Scientific™
Product Code. 15480507
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480507 5 g 5g
15490507 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15480507 Supplier Alfa Aesar™ Supplier No. J66159.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
A α-anomeric form of glucose which contains a phosphate group on the primary carbon. Used as a substance for the estimation of phosphorylases.
Solubility
Soluble in water
Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 1233690-06-6
Molecular Formula C6H17K2O11P
Molecular Weight (g/mol) 374.361
MDL Number MFCD00149435
InChI Key RZULLROGARPECB-QMKHLHGBSA-N
Synonym alpha-d-glucopyranose, 1-dihydrogen phosphate , dipotassium salt, dihydrate, alpha-d-glucopyranose 1-phosphate dipotassium salt hydrate, alpha-d-glucose 1-phosphate dipotassium salt dihydrate, alpha-glucose-1-phosphate dipotassium dihydrate, alpha-glucose-1-phosphate dipotassium salt dihydrate mi, a-d-glucose-1-phosphate dipotassium salt hydrate, alpha-d-glucose-1-phosphate dipotassium salt hydrate, dipotassium dihydrate ?-d-glucopyranosyl phosphate, dipotassium 2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl phosphate dihydrate, potassium 2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl phosphate dihydrate
PubChem CID 131855785
IUPAC Name potassium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate;dihydrate
SMILES C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.O.O.[K].[K]
Spezifikation
Color White
Odor Odorless
Quantity 5 g
Merck Index 14,4461
Solubility Information Soluble in water
Formula Weight 336.32 (Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material alpha-D-Glucose-1-phosphate dipotassium salt hydrate