missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyano-4-methoxycinnamic acid, 98+%, Thermo Scientific™

Product Code. 15408839
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408839 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15408839 Supplier Thermo Scientific Alfa Aesar Supplier No. L05299.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1519-55-7
Melting Point ∽228°C (decomposition)
Molecular Formula C11H9NO3
MDL Number MFCD00051722
Quantity 5 g
Beilstein 2110241
Synonym 2z-2-cyano-3-4-methoxyphenyl prop-2-enoic acid, 2-cyano-3-4-methoxyphenyl acrylic acid, alpha-cyano-4-methoxycinnamic acid, z-2-cyano-3-4-methoxyphenyl prop-2-enoic acid
InChI Key KMHNRJDDHTZZDZ-TWGQIWQCSA-N
SMILES COC1=CC=C(\C=C(\C#N)C(O)=O)C=C1
IUPAC Name (E)-2-cyano-3-(4-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 203.20
PubChem CID 736236
Formula Weight 203.2
Percent Purity ≥98%
Chemical Name or Material alpha-Cyano-4-methoxycinnamic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.