Learn More
Invitrogen™ Alpha-Bungarotoxin Conjugates
Product Code. 17806578
Change view
Click to view available options
Label or Dye:
Alexa Fluor™ 488
Alexa Fluor™ 555
Alexa Fluor™ 594
Alexa Fluor™ 647
Alexa Fluor™ Plus 405
Biotin-XX
Tetramethylrhodamine
Unlabeled
Unit Size:
100µL
1mg
500µg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Label or Dye unitSize
17806578 Alexa Fluor™ Plus 405 100µL
11554147 Alexa Fluor™ 488 500µg
11554187 Alexa Fluor™ 555 500µg
11564147 Alexa Fluor™ 594 500µg
11544187 Alexa Fluor™ 647 500µg
11534147 Biotin-XX 500µg
11500756 Tetramethylrhodamine 500µg
10729564 Unlabeled 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17806578 Supplier Invitrogen™ Supplier No. B56130
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates.
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions.
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions. Our bright and photostable Alexa Fluor 555 alpha-bungarotoxin is a superior choice of probe for visualizing this receptor.
Fluorescent alpha-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and localization of neuromuscular junctions. Conjugated alpha-bungarotoxin has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).
TRUSTED_SUSTAINABILITY
Specifications
Color Violet
Quantity 100 μg
Content And Storage Store at ≤-20°C, protect from light. Avoid freeze-thaw cycles.
Label or Dye Alexa Fluor™ Plus 405
Excitation/Emission 411/431 nm
Product Line Alexa Fluor