missing translation for 'onlineSavingsMsg'
Learn More

Aloin B, MedChemExpress

Product Code. 16411578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411578 5 mg 5mg
16409168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16411578 Supplier Medchem Express Supplier No. HYN0886
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Aloin B is one isomer of Aloin; Aloin is a physiologically active anthraquinone present in aloe.

Aloin B is one isomer of Aloin

Specifications

Chemical Name or Material Aloin B
CAS 28371-16-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22O9
Quantity 5 mg
Synonym Aloin-B Isobarbaloin
Solubility Information DMSO : 50 mg/mL (119.51 mM; Need ultrasonic)
SMILES O[C@H]([C@H]1O)[C@](O[C@H](CO)[C@H]1O)([H])[C@@](C2=CC=CC(O)=C23)([H])C(C=C(CO)C=C4O)=C4C3=O
Molecular Weight (g/mol) 418.39
Formula Weight 418.39
Percent Purity 99.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.