missing translation for 'onlineSavingsMsg'
Learn More

Aloin B, Dr. Ehrenstorfer

Product Code. 30320070
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320070 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320070 Supplier Dr. Ehrenstorfer Supplier No. DREC10142100
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aloin B
CAS 28371-16-6
Analyte or Component Names Aloin B
Shipping Condition +20°C
Molecular Formula C21 H22 O9
Certifications/Compliance ISO/IEC 17025
Synonym 9(10H)-Anthracenone, 10-β-D-glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-, (10R)-, (10H)-Anthracenone, 10-β-D-glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-, (R)-, Anthrone, 10-glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-, stereoisomer (8CI), Isobarbaloin (7CI), (10R)-10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone, 10-epi-Aloin A, Aloin, Aloin B
InChI Formula InChI=1S/C21H22O9/c22-6-8-4-10-14(21-20(29)19(28)17(26)13(7-23)30-21)9-2-1-3-11(24)15(9)18(27)16(10)12(25)5-8/h1-5,13-14,17,19-26,28-29H,6-7H2/t13-,14-,17-,19+,20-,21+/m1/s1
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]2c3cccc(O)c3C(=O)c4c(O)cc(CO)cc24
IUPAC Name (10R)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-10H-anthracen-9-one
Molecular Weight (g/mol) 418.39
Quantity 10 mg
Formula Weight 418.1264 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.