missing translation for 'onlineSavingsMsg'
Learn More

Aloe-emodin, TRC

Product Code. 17923275
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17923275 100 mg 100mg
17982775 50 mg 50mg
17931255 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17923275 Supplier Toronto Research Chemicals Supplier No. TRCA575400100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 481-72-1
Molecular Formula C15 H10 O5
Molecular Weight (g/mol) 270.24
Synonym 1,8-Dihydroxy-3-(hydroxymethyl)-9,10-anthracenedione, 1,8-Dihydroxy-3-hydroxymethylanthraquinone, 3-Hydroxymethylchrysazin, 3-Hydroxymethylchrysazine, Aloe emodin, Aloe-emodol, NSC 38628, Rhabarberone
IUPAC Name 1,8-dihydroxy-3-(hydroxymethyl)anthracene-9,10-dione
SMILES OCc1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H10O5/c16-6-7-4-9-13(11(18)5-7)15(20)12-8(14(9)19)2-1-3-10(12)17/h1-5,16-18H,6H2
Formula Weight 270.0528
Percent Purity >95%
Chemical Name or Material Aloe Emodin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.