missing translation for 'onlineSavingsMsg'
Learn More

Allyl Ester of Atorvastatin Cyclic (Isopropyl) Impurity, TRC

Product Code. 30334201
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30334201 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334201 Supplier Toronto Research Chemicals Supplier No. TRCA79183510MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allyl Ester of Atorvastatin Cyclic (Isopropyl) Impurity
CAS 1316295-72-3
Molecular Formula C36H39FN2O7
Quantity 10 mg
Synonym 7-(4-Fluorophenyl)hexahydro-beta,7-dihydroxy-1b-(1-methylethyl)-7a-phenyl-1a-[(phenylamino)carbonyl]-3H-oxireno[3,4]pyrrolo[2,1-b][1,3]oxazine-3-butanoic Acid Allyl Ester, Allyl-ATV-cycloIP
InChI Formula InChI=1S/C36H39FN2O7/c1-4-21-44-31(41)23-29(40)22-30-19-20-39-35(43,26-15-17-27(37)18-16-26)33(25-11-7-5-8-12-25)34(46-33,36(39,45-30)24(2)3)32(42)38-28-13-9-6-10-14-28/h4-18,24,29-30,40,43H,1,19-23H2,2-3H3,(H,38,42)
SMILES CC(C)C12OC(CC(O)CC(=O)OCC=C)CCN1C(O)(c3ccc(F)cc3)C4(OC24C(=O)Nc5ccccc5)c6ccccc6
IUPAC Name prop-2-enyl 4-[7-(4-fluorophenyl)-7-hydroxy-7a-phenyl-1a-(phenylcarbamoyl)-1b-propan-2-yl-4,5-dihydro-3H-oxireno[1,2]pyrrolo[4,5-b][1,3]oxazin-3-yl]-3-hydroxybutanoate
Molecular Weight (g/mol) 630.7
Formula Weight 630.2741
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.