Learn More
Allyl-d5 Bromide, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Allyl-d5 Bromide |
| CAS | 102910-37-2 |
| Molecular Formula | C3 2H5 Br |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C3H5Br/c1-2-3-4/h2H,1,3H2/i1D2,2 D,3D2 |
| SMILES | [2 H]C(=C([2 H])C([2 H])([2 H])Br)[2 H] |
| IUPAC Name | 3-bromo-1,1,2,3,3-pentadeuterioprop-1-ene |
| Molecular Weight (g/mol) | 126.01 |
| Formula Weight | 124.9888 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |