missing translation for 'onlineSavingsMsg'
Learn More

Allotetrahydrocorticosterone, TRC

Product Code. 30133993
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133993 100 mg 100mg
30134949 50 mg 50mg
30005308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133993 Supplier Toronto Research Chemicals Supplier No. TRCA548495100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-63-5
Molecular Formula C21H34O4
Molecular Weight (g/mol) 350.49
Synonym (3alpha,5alpha,11beta)-3,11,21-Trihydroxypregnan-20-one, 3alpha,11beta,21-Trihydroxy-5alpha-pregnan-20-one, 3alpha,5alpha-Tetrahydrocorticosterone, 5alpha-Pregnane-3alpha,11beta,21-triol-20-one, 5alpha-THB, 5alpha-Tetrahydrocorticosterone, 3alpha,11beta,21-Trihydroxy-5alpha-pregnan-20-one;
IUPAC Name 1-[(3R,5S,8S,9S,10S,11S,13S,14S,17S)-3,11-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-hydroxyethanone
SMILES C[C@]12CC[C@@H](O)C[C@@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H34O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h12-17,19,22-24H,3-11H2,1-2H3/t12-,13+,14-,15-,16+,17-,19+,20-,21-/m0/s1
Formula Weight 350.2457
Percent Purity >95%
Chemical Name or Material Allotetrahydrocorticosterone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.