CAS RN 600-63-5
CAS RN 600-63-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Allotetrahydrocorticosterone, TRC
CAS: 600-63-5 Molecular Formula: C21H34O4 Molecular Weight (g/mol): 350.49 Synonym: (3alpha,5alpha,11beta)-3,11,21-Trihydroxypregnan-20-one,3alpha,11beta,21-Trihydroxy-5alpha-pregnan-20-one,3alpha,5alpha-Tetrahydrocorticosterone,5alpha-Pregnane-3alpha,11beta,21-triol-20-one,5alpha-THB,5alpha-Tetrahydrocorticosterone,3alpha,11beta,21-Trihydroxy-5alpha-pregnan-20-one; IUPAC Name: 1-[(3R,5S,8S,9S,10S,11S,13S,14S,17S)-3,11-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-hydroxyethanone SMILES: C[C@]12CC[C@@H](O)C[C@@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23