Learn More
Allocholic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Allocholic Acid |
| CAS | 2464-18-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H40 O5 |
| Quantity | 5 mg |
| Synonym | 3-α,7-α,12-α-Trihydroxy-5-α-cholanoic Acid, 5α-Cholic Acid |
| InChI Formula | InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14-,15-,16-,17+,18+,19-,20+,22+,23+,24-/m1/s1 |
| SMILES | C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| IUPAC Name | (4R)-4-[(3R,5R,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
| Molecular Weight (g/mol) | 408.57 |
| Show More |