missing translation for 'onlineSavingsMsg'
Learn More

all-trans-4-Oxoretinoic acid, MedChemExpress

Product Code. 16496837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16496837 Supplier Medchem Express Supplier No. HY107494A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress all-trans-4-Oxoretinoic acid, an active metabolite of vitamin A, induces gene transcription via binding to nuclear retinoic acid receptors (RARs).

all-trans-4-Oxoretinoic acid, an active metabolite of vitamin A, induces gene transcription via binding to nuclear retinoic acid receptors (RARs).

Specifications

Chemical Name or Material all-trans-4-Oxoretinoic acid
CAS 38030-57-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H26O3
Quantity 5 mg
Synonym all-trans 4-Keto Retinoic Acid
Solubility Information DMSO : 50 mg/mL (159.02 mM; Need ultrasonic)
SMILES CC(/C=C/C=C(/C=C/C1=C(C(CCC(C)1C)=O)C)C)=C\C(O)=O
Molecular Weight (g/mol) 314.42
Formula Weight 314.42
Percent Purity 90.03%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.