Learn More
all-trans 4-Keto Retinoic Acid, TRC
Product Code. 30137688
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137688 5 mg 5mg
30133977 2.5 mg 2.5mg
30011819 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137688 Supplier Toronto Research Chemicals Supplier No. TRCK2049805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38030-57-8
Molecular Formula C20 H26 O3
Molecular Weight (g/mol) 314.42
Synonym BAL10084, GSK1044133A, BAL0010084-000, BAL0004079-000, BAL4079, 9-cis retinoic acid, alitretinoin related
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES C\C(=C/C=C/C(=C/C(=O)O)/C)\C=C\C1=C(C)C(=O)CCC1(C)C
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C20H26O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7+,15-13+
Formula Weight 314.1882
Percent Purity >95%
Chemical Name or Material all-trans 4-Keto Retinoic Acid
Grade HPLC