Learn More
Cytiva AlkPhos Direct™ Hybridization Buffer for Direct Labeling System
Product Code. 10566875 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
For 5000 cm2 membrane
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10566875 For 5000 cm2 membrane Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10566875 Supplier Cytiva Supplier No. RPN3688
  • ONLINE EXCLUSIVE PRICE
    913.01 EUR
    766.24 EUR / Pack of 1
    Save 146.77 EUR
    16% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Includes
Each system includes the following reagents, sufficient for 25 or 50 labeling reactions and detection of 5000cm2 membrane: labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star™ or ECF detection reagents.
Fast and convenient system for all high-sensitivity applications using DNA, RNA, or oligonucleotide probes
  • Hybridization stringency can be controlled by elevating temperatures up to 75°C (167°F), or by varying salt concentration
  • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
  • ECF™ detection for quantitation experiments
  • Thirty minutes from completion of hybridization to application of detection reagent: saves two to three hours over conventional indirect methods
  • Resulting probes can be used without purification
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Fast, convenient for high-sensitivity applications with DNA, RNA, Oligo probes
Includes Labeling reagent, crosslinker, control DNA, Reaction Buffer, hybridization Buffer, blocking agent, CDP-Star™ or ECF detection reagents
Product Type Amersham AlkPhos Direct Hybridization Buffer sufficient for 5000 cm2
Quantity For 5000 cm2 membrane